Learn More
ACROBiosystems Recombinant Protein;Mouse IL-4 R alpha / CD124 Protein, Fc Tag;HEK293;1MG;ILR,IL4R,IL-4R,IL4 R,CD124,IL-4RA,IL4RA,Interleukin-4 receptor,Interleukin 4 receptor,IL-4 R alpha
Supplier:  ACROBiosystems ILRM52521MG
Encompass_Preferred
Recombinant Protein;Mouse IL-4 R alpha / CD124 Protein, Fc Tag;1MG;Mouse IL-4 R alpha, Fc Tag (ILR-M5252) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Arg 233 (Accession # NP_001008700).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8710