Learn More
ACROBiosystems Recombinant Protein;Human IL-4 R alpha / CD124 Protein;HEK293;50UG;ILR,IL4R,IL-4R,IL4 R,CD124,IL-4RA,IL4RA,Interleukin-4 receptor,Interleukin 4 receptor,IL-4 R alpha
Catalog No. 501657650
Encompass_Preferred
Recombinant Protein;Human IL-4 R alpha / CD124 Protein;50UG;Human IL-4 R alpha, His Tag (ILR-H5221) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession # NP_000409.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.5 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-4 R alpha, His Tag (Cat. No. ILR-H5221) at 5μg/mL (100 μL/well) can bind Biotinylated Human IL-4, His Tag (Cat. No. IL4-H82E0) with a linear range of 1.95-15.6 ng/mL (QC tested).
Catalog No. 50-165-7650 Supplier ACROBiosystems Supplier No. ILRH522150UG
Recombinant Protein;Human IL-4 R alpha / CD124 Protein;50UG;Human IL-4 R alpha, His Tag (ILR-H5221) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession # NP_000409.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.5 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-4 R alpha, His Tag (Cat. No. ILR-H5221) at 5μg/mL (100 μL/well) can bind Biotinylated Human IL-4, His Tag (Cat. No. IL4-H82E0) with a linear range of 1.95-15.6 ng/mL (QC tested).