Learn More
Aviva Systems Biology Corp IL5 Antibody - middle region (ARP54360_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54360_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IL5. Alternate names: EDF, IL-5, TRFApplicationWB. Species reactivity: Human
Catalog No. 50-014-61185