Learn More
Aviva Systems Biology Corp Il5 Antibody - middle region (ARP54359_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54359_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Il5. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Il-5ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-912