Learn More
Sino Biological IL-5/IL5 / Interleukin 5 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 15673R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-5/IL5 / Interleukin 5 (rh IL-5/IL5 / Interleukin 5; Catalog#15673-HNCE; P05113; Ile20-Ser134).
Catalog No. 50-224-7690