Learn More
Sino Biological IL-5/IL5/Interleukin 5 Neutralizing Antibody (HRP) 100ug
Catalog No. 502248233
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-5/IL5/Interleukin 5 (rh L-5/IL5/Interleukin 5; Ile20-Ser134; P05113; Catalog#15673-HNCE)and conjugated with horseradish-peroxidase (HRP).
This item is not returnable. View return policy
Catalog No. 50-224-8233 Supplier Sino Biological Supplier No. 15673R013H100
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-5/IL5/Interleukin 5 (rh L-5/IL5/Interleukin 5; Ile20-Ser134; P05113; Catalog#15673-HNCE)and conjugated with horseradish-peroxidase (HRP).