Learn More
ACROBiosystems Recombinant Protein;Human IL-5 R alpha / CD125 Protein;HEK293;50UG;ILA,IL5RA,CD125,IL-5RA,IL-5R-alpha,CDw125,IL5R
Catalog No. 501658800
Encompass_Preferred
Recombinant Protein;Human IL-5 R alpha / CD125 Protein;50UG;Human IL-5 R alpha, His Tag (ILA-H52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 21 - Glu 335 (Accession # NP_000555).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.7 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8800 Supplier ACROBiosystems Supplier No. ILAH52H450UG
Recombinant Protein;Human IL-5 R alpha / CD125 Protein;50UG;Human IL-5 R alpha, His Tag (ILA-H52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 21 - Glu 335 (Accession # NP_000555).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.7 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.