Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3569RTM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: IL6/1270. Clonality: Monoclonal. Host: Rat. Isotype: IgG1 kappa. Positive Control: Heart. Cellular Staining: Cytoplasmic, extracellular (secreted). Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant IL-6 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 21kDa.Chromosome: 7p21. Gene Symbol: IL6. Gene ID: 3569. Ensemble: ENSG00000136244. Uniprot: NP_000591.
Catalog No. 50-188-4828