Learn More
Sino Biological IL-6 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 80076R007100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-6 (rR IL-6; Catalog#80076-RNAE; NP_036721.1; Phe25-Thr211).
Catalog No. 50-224-7348