Learn More
Enzo Life Sciences IL-6 receptor Recombinant monoclonal antibody (rhPM-1) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5500200
Encompass_Preferred
Clone: rhPM-1 (Tocilizumab). Isotype: IgGκ. Immunogen: This antibody was prepared by CDR grafting of mouse AUK 12-20 antibody onto a human framework regions. Species reactivity: Human. Specificity: Recognizes both the membrane bound and soluble forms of human IL-6R. Applications: ELISA. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3076