Learn More
ACROBiosystems Recombinant Protein;Human IL-6 R alpha / CD126 Protein;HEK293;1MG;ILR,IL6R,IL6 R,IL-6R,CD126,IL-6R-1,IL-6RA,IL6RA,IL-6R-alpha,IL6RQ,gp80,IL-6 R alpha,IL-6Ra
Supplier:  ACROBiosystems ILRH42231MG
Encompass_Preferred
Recombinant Protein;Human IL-6 R alpha / CD126 Protein;1MG;Human IL-6 R alpha, His Tag (ILR-H4223) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 365 (Accession # NP_000556.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40 kDa. The protein migrates as 55-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-6, Tag Free (Cat. No. IL6-H4218) at 5 μg/mL (100 μL/well) can bind Human IL-6 R alpha, His Tag (Cat. No. ILR-H4223) with a linear range of 1-16 ng/mL (QC tested).
Catalog No. 50-165-7656