Learn More
ACROBiosystems Human IL-6 R Alpha Protein, His Tag, 50 μg
Catalog No. 501657655
Encompass_Preferred
Human IL-6 R alpha/CD126 is a pleiotropic cytokine receptor subunit involved in immune response and acute phase reactions. This recombinant protein is expressed in HEK293 cells and contains amino acids Leu 20 to Pro 365, with a C-terminal polyhistidine tag. It is characterized by high purity and verified by mass spectrometry and SEC-MALS.
  • Greater than 95% purity as determined by SDS-PAGE and SEC-MALS.
  • Calculated molecular weight of 39.4 kDa, migrating as 55-75 kDa under reducing conditions due to glycosylation.
  • Endotoxin level less than 0.01 EU per μg.
  • Lyophilized from a 0.22 μm filtered PBS solution, pH 7.4, with trehalose.
  • 100% sequence coverage verified by LC-MS/MS peptide mapping.
  • Validated biological activity with a BLI affinity constant of 20.3 nM.
  • Stable for 12 months at -20°C to -70°C in lyophilized state.
Catalog No. 50-165-7655 Supplier ACROBiosystems Supplier No. ILRH422350UG
Human IL-6 R alpha/CD126 is a pleiotropic cytokine receptor subunit involved in immune response and acute phase reactions. This recombinant protein is expressed in HEK293 cells and contains amino acids Leu 20 to Pro 365, with a C-terminal polyhistidine tag. It is characterized by high purity and verified by mass spectrometry and SEC-MALS.
  • Greater than 95% purity as determined by SDS-PAGE and SEC-MALS.
  • Calculated molecular weight of 39.4 kDa, migrating as 55-75 kDa under reducing conditions due to glycosylation.
  • Endotoxin level less than 0.01 EU per μg.
  • Lyophilized from a 0.22 μm filtered PBS solution, pH 7.4, with trehalose.
  • 100% sequence coverage verified by LC-MS/MS peptide mapping.
  • Validated biological activity with a BLI affinity constant of 20.3 nM.
  • Stable for 12 months at -20°C to -70°C in lyophilized state.