Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human IL-6 R alpha / CD126 Protein, Avi Tag (Avitag™);HEK293;200UG;CD6,IL6R,IL6 R,IL-6R,CD126,IL-6R-1,IL-6RA,IL6RA,IL-6R-alpha,IL6RQ,gp80,IL-6 R alpha,IL-6Ra
Supplier:  ACROBiosystems CD6H82E8200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human IL-6 R alpha / CD126 Protein, Avi Tag (Avitag™);200UG;Biotinylated Human IL-6 R alpha, His Tag (CD6-H82E8) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 365 (Accession # P08887-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 41.7 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-6, Tag Free (Cat. No. IL6-H4218) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-6 R alpha, His Tag (Cat. No. CD6-H82E8) with a linear range of 4-125 ng/mL (QC tested).
Catalog No. 50-165-8395