Learn More
Sino Biological IL6ST/gp130/CD130 Antibody (PE), Mouse MAb 100Tests
Supplier:  Sino Biological 10974MM11P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL6ST/gp130/CD130 (rh IL6ST/gp130/CD130; Catalog#10974-HCCH; NP_002175.2; Met1-Ile618) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-224-7600