Learn More
Eurofins Discoverx PathHunter IL7R/2RG Bioassay Kit, 10-Plate. (93-0997Y13-00082)
Catalog No. 502001785
Encompass
The PathHunter IL7R/IL2RG Dimerization Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. IL-7) induced dimerization of IL7R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL7R target protein fused to PK alongside IL2RG target protein fused to EA. Upon ligand-induced dimerization of IL7R/IL2RG, detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-1785 Supplier Eurofins Discoverx Supplier No. 930997Y1300082
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL7R/IL2RG Dimerization Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. IL-7) induced dimerization of IL7R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL7R target protein fused to PK alongside IL2RG target protein fused to EA. Upon ligand-induced dimerization of IL7R/IL2RG, detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.