Learn More
Aviva Systems Biology Corp IL7R Antibody - middle region (ARP59084_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59084_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL7R. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human IL7R. Alternate names: CD127, CDW127, IL-7R-alpha, IL7RA, ILRAApplicationWB. Species reactivity: Dog, Horse, Human
Catalog No. 50-286-572