Learn More
Enzo Life Sciences IL-7 receptor alpha (human) monoclonal antibody (R34-34) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2530100
Encompass_Preferred
Clone: R34-34. Isotype: IgG1. Immunogen: Human leukemic pre-B cells. Species reactivity: Human Dog. Applications: Flow Cytometry, IP, FUNC. Formulation: Liquid. In TRIS-NaCl, pH 8.
Catalog No. 50-201-2799