Learn More
Enzo Life Sciences IL-7 receptor alpha (human) monoclonal antibody (R34-34) (Alexa Fluor 647) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2510100
Encompass_Preferred
Clone: R34-34. Isotype: IgG1. Immunogen: Human leukemic pre-B cells. Species reactivity: Human Dog. Applications: Flow Cytometry. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-201-2797