Learn More
ACROBiosystems Recombinant Protein;ActiveMax Recombinant Human IL-7;HEK293;50UG;IL7,IL7,IL-7,Interleukin 7,Interleukin-7,Interleukin7
Supplier:  ACROBiosystems IL7H421950UG
Encompass_Preferred
Recombinant Protein;ActiveMax Recombinant Human IL-7;50UG;Human IL-7, Tag Free (IL7-H4219) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession # P13232).;This protein carries no "tag". The protein has a calculated MW of 17.4 kDa. The protein migrates as 24 kDa and 25-28 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human IL-7, Tag Free (Cat. No. IL7-H4219) at 2 g/mL (100 L/well) can bind Human IL-7 R alpha, Fc Tag (Cat. No. ILA-H5258) with a linear range of 1-16 ng/mL (QC tested).;ActiveMaxHumanIL-7, Tag Free(Cat. No. IL7-H4219)stimulates proliferationof PHA-P-activatedhuman peripheralblood mononuclearcell (PBMC).TheED50for this effect is 0.108-0.829 ng/ml (Routinely tested).
Catalog No. 50-165-7659