Learn More
Aviva Systems Biology Corp IL8RB Antibody - N-terminal region (ARP41956_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41956_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL8RB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IL8RB. Alternate names: CD182, CDw128b, CMKAR2, CXCR2, IL8R2, IL8RA, IL8RBApplicationWB. Species reactivity: Human
Catalog No. 50-014-58063