Learn More
Medchemexpress LLC HY-101664 5mg Medchemexpress, Ilaprazole CAS:172152-36-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1016645MG
Encompass_Preferred
Medchemexpress, HY-101664 5mg Ilaprazole CAS:172152-36-2 Ilaprazole (IY-81149) is a proton pump inhibitor; inhibits H + /K + -ATPase with an IC 50 of 6.0 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2191