Learn More
Medchemexpress LLC Ilaprazole-d3 | 1070969-17-3 | 98.0% | 369.46 | C19H15D3N4O2S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY101664S1MG
Encompass_Preferred
Ilaprazole-d3 is a deuterium-labeled analogue of ilaprazole intended for use as a tracer and internal standard in analytical research. Supplied in small quantities for laboratory use, it supports accurate quantification in metabolic and pharmacokinetic studies using MS and NMR techniques.
  • Deuterium-labeled analogue for isotope tracing.
  • Suitable as an internal standard for quantitative LC-MS, GC-MS, and NMR.
  • Molecular formula C19H15D3N4O2S; molecular weight 369.46 g·mol⁻¹.
  • Provided in small, research-scale quantities for analytical workflows.
  • Refer to certificate of analysis for lot-specific purity and handling information.
Catalog No. 50-002-45448