Learn More
Medchemexpress LLC Ilaprazole sodium hydrate | 2322264-11-7 | 99.15% | 250 MG
SDP
Catalog No. 5000414438
Encompass_Preferred
Ilaprazole (IY-81149) sodium hydrate is an orally active proton pump inhibitor. It irreversibly inhibits H+/K+-ATPase in a dose-dependent manner with an IC50 of 6 μM in rabbit parietal cell preparation. It is used for the research of gastric ulcers and is also a potent TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor.
  • Orally active proton pump inhibitor
  • Irreversibly inhibits H+/K+-ATPase
  • Used for research of gastric ulcers
  • Potent TOPK inhibitor
Catalog No. 50-004-14438 Supplier Medchemexpress LLC Supplier No. 250MGHYB2145A
Ilaprazole (IY-81149) sodium hydrate is an orally active proton pump inhibitor. It irreversibly inhibits H+/K+-ATPase in a dose-dependent manner with an IC50 of 6 μM in rabbit parietal cell preparation. It is used for the research of gastric ulcers and is also a potent TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor.
  • Orally active proton pump inhibitor
  • Irreversibly inhibits H+/K+-ATPase
  • Used for research of gastric ulcers
  • Potent TOPK inhibitor