Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILC Dover Sentinel XL HP Half Hood

Catalog No. 19631044
Change view
Click to view available options
For Use With (Equipment):
Sentinel XL HP system, CBRN system
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-631-044 Sentinel XL HP system, CBRN system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-631-044 Supplier ILC Dover Supplier No. S40013
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Sentinel XL half hood head cover offers an APF of 25, provide a wide field of view and easily accommodates stethoscopes.

  • Offers an APF of 25, provide a wide field of view and easily accommodates stethoscopes

Specifications

Color White
Material Polyethylene, Polyolefin
Size One Size
For Use With (Equipment) Sentinel XL HP system, CBRN system
Includes 3 head covers
Material (Lens) PET; Polyester Film
Quantity 3/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.