Learn More
ILC Dover SCape CO/CBRN 30 Escape Respirator
Catalog No. 19631055
Change view
Click to view available options
For Use With (Application):
Provides smoke protection
Catalog No. For Use With (Application)
19-631-055 Provides smoke protection
1 options
Catalog No. 19-631-055 Supplier ILC Dover Supplier No. 60001043503
Add to Cart
Add to Cart
SCape™ CO/CBRN30 from ILC Dover provides mission-critical protection from carbon monoxide, chemical, biological, radiological and nuclear contaminants.
  • Provides mission-critical protection from carbon monoxide, chemical, biological, radiological and nuclear contaminants
Compliance
NIOSH Approved TC-14G-0277/TC-146.0312
Specifications
Certifications/Compliance NIOSH
Product Type Escape Hood
For Use With (Application) Provides smoke protection
Includes Integral 64 lmp blower with indicator light, exhaust valve, one-size neck seal, military grade filter canister, hepa filter, multi layer nylon film lens, battery, clear hood with large 360 vision
Material Polyethylene
Quantity 1 Ea.
Size Universal