Learn More
ILC Dover Sentinel XL™ CBRN PAPR
Description
Unique lay flat hood design saves space during storage and allows the hood to fit into a customized NIOSH approved backpack
- Durable dual chamber blower designed for backpack or belt mounting
- Butyl hood includes a comfortable, adjustable head band
- Sentinel is the only NIOSH approved CAP 2 loose fitting CBRN PAPR—delivers twice the filter performance of a CAP 1 system
- Butyl hood includes a comfortable, adjustable head band
Components
- Blower with battery
- 3 CBRN filters
- Lay flat butyl hood
- Battery charger (for rechargeable systems)
- Flow meter
- User's manual
- Waist belt
Order Info
Available in Lithium battery assembly
Specifications
Specifications
| Product Type | PAPR System with Hood |
| Size | One Size |
| For Use With (Application) | Ultimate in chemical and biological protection |
| Includes | Cbrn duffle |
| Model | S-5000-04 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.