Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ile-AMS TFA | 219931-45-0 | MFCD31619268 | 98.6% | 572.52 g/mol | C18H27F3N8O8S | 10 MG
SDP

Catalog No. 5000288298
Encompass_Preferred

Ile-AMS TFA is the trifluoroacetic acid (TFA) salt of an aminoacyl-sulfamoyl adenosine analogue that inhibits isoleucyl-tRNA synthetase and displays potent antiparasitic activity (P. falciparum ABS IC50 = 1.19 nM). Intended for laboratory research use only.

  • Purity 98.55%.
  • Molecular weight 572.52 g/mol; chemical formula C18H27F3N8O8S.
  • Soluble in water (≈100 mg/mL); ultrasonic assistance may be required.
  • Recommended storage: sealed, away from moisture; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research quantities, including a 10 mg package.
  • Used as a tool compound for studies of aminoacyl-tRNA synthetase inhibition and antiparasitic research.

Catalog No. 50-002-88298 Supplier Medchemexpress LLC Supplier No. HY156895A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ile-AMS TFA is the trifluoroacetic acid (TFA) salt of an aminoacyl-sulfamoyl adenosine analogue that inhibits isoleucyl-tRNA synthetase and displays potent antiparasitic activity (P. falciparum ABS IC50 = 1.19 nM). Intended for laboratory research use only.

  • Purity 98.55%.
  • Molecular weight 572.52 g/mol; chemical formula C18H27F3N8O8S.
  • Soluble in water (≈100 mg/mL); ultrasonic assistance may be required.
  • Recommended storage: sealed, away from moisture; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research quantities, including a 10 mg package.
  • Used as a tool compound for studies of aminoacyl-tRNA synthetase inhibition and antiparasitic research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.