Learn More
CPC Scientific H-Ile-Thr-Asp-Gln-Val-Pro-Phe-Ser-Val-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific CANS001A
Encompass
A melanogenic enzyme that is recognized in the context of HLA-A2 by tumo-rinfiltrating lymphocytes (TIL) in vivo. ONE-LETTER SEQUENCE: ITDQVPFSVMOLECULAR FORMULA: C46H72N10O15MOLECULAR WEIGHT:1005.14STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [162558-10-3], SYNONYMS: Melanocyte Protein PMEL 17 (185-193) (human, bovine, mouse), ME20M/ME20S (130-138) (human), Melanoma-Associated ME20 Antigen (185-193) (human, bovine, mouse), Melanocyte Lineage-Specific Antigen GP100 (185-193) (human, bovine, mouse), ME20-M/ME20-S (130-138) (human), 95 kDa Melanocyte-Specific Secreted Glycoprotein (130-138) (human)RESEARCH AREA: Cancer
Catalog No. 50-210-8901