Learn More
Bachem (Ile8)-Oxytocin Trifluoroacetate
SDP
Supplier:  Bachem 4030888.0005
Encompass_Preferred
(Ile8)-Oxytocin Trifluoroacetate, H-Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Ile-Gly-NH2, 362-39-0, C43H66N12O12S2, Mr 1007.20, 5mg. Mesotocin. Mesotocin is an oxytocin analog produced in birds, reptiles, and amphibians as well as in most marsupials.
Catalog No. 50-259-614