Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ilexhainanoside D | 1137648-52-2 | 664.82 | C36H56O11 | 10 MG
SDP

Catalog No. 5000289763
Encompass_Preferred

Ilexhainanoside D is a triterpenoid saponin extracted from Ilex hainanensis Merr. It is used in research for its reported anti-inflammatory activity and is supplied as a purified research compound (CAS 1137648-52-2). Handle and store under recommended conditions to preserve stability.

  • Triterpenoid saponin isolated from Ilex hainanensis
  • Reported anti-inflammatory activity in combination with ilexsaponin A1
  • Molecular formula C36H56O11; molecular weight 664.82
  • Store at 4°C; in solvent: -80°C for up to 6 months, -20°C for up to 1 month; protect from light
  • Supplied in small research-scale quantities for laboratory use

Catalog No. 50-002-89763 Supplier Medchemexpress LLC Supplier No. HYN504010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ilexhainanoside D is a triterpenoid saponin extracted from Ilex hainanensis Merr. It is used in research for its reported anti-inflammatory activity and is supplied as a purified research compound (CAS 1137648-52-2). Handle and store under recommended conditions to preserve stability.

  • Triterpenoid saponin isolated from Ilex hainanensis
  • Reported anti-inflammatory activity in combination with ilexsaponin A1
  • Molecular formula C36H56O11; molecular weight 664.82
  • Store at 4°C; in solvent: -80°C for up to 6 months, -20°C for up to 1 month; protect from light
  • Supplied in small research-scale quantities for laboratory use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.