Learn More
Medchemexpress LLC Ilexsaponin A | 108524-93-2 | 1 ML
SDP
Catalog No. 5000421422
Encompass_Preferred
Ilexsaponin A, isolated from the root of Ilex pubescens, is a chemical compound that attenuates ischemia-reperfusion-induced myocardial injury through an anti-apoptotic pathway. It is intended for research and analytical applications, offering a reliable tool for studying cellular processes.
  • Attenuates ischemia-reperfusion-induced myocardial injury
  • Functions via an anti-apoptotic pathway
  • Reduces myocardial infarct size
  • Lowers serum levels of LDH, AST, and CK-MB
  • Increases cellular viability
  • Inhibits apoptosis in hypoxia/reoxygenation cardiomyocytes
  • Offers a purity of 99.53%
Catalog No. 50-004-21422 Supplier Medchemexpress LLC Supplier No. HYN263810MM/1ML
Ilexsaponin A, isolated from the root of Ilex pubescens, is a chemical compound that attenuates ischemia-reperfusion-induced myocardial injury through an anti-apoptotic pathway. It is intended for research and analytical applications, offering a reliable tool for studying cellular processes.
  • Attenuates ischemia-reperfusion-induced myocardial injury
  • Functions via an anti-apoptotic pathway
  • Reduces myocardial infarct size
  • Lowers serum levels of LDH, AST, and CK-MB
  • Increases cellular viability
  • Inhibits apoptosis in hypoxia/reoxygenation cardiomyocytes
  • Offers a purity of 99.53%