Learn More
Aviva Systems Biology Corp ILF3 Antibody - C-terminal region (ARP38969_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38969_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ILF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ILF3. Alternate names: CBTF, DRBF, MMP4, MPP4, NF90, NFAR, NF110, NF90a, NF90b, NFAR2, TCP80, DRBP76, NF110b, NFAR-1, TCP110, MPHOSPH4, NF-AT-90ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56873