Learn More
Aviva Systems Biology Corp ILF3 Antibody - C-terminal region (ARP32666_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32666_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ILF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ILF3. Alternate names: DRBF, DRBP76, MMP4, MPHOSPH4, MPP4, NF-AT-90, NF90, NFAR, NFAR-1, TCP80, CBTF, NF110, NF90a, NF90b, NFAR2, NF110b, TCP110ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54697