Learn More
Aviva Systems Biology Corp ILF3 Antibody - N-terminal region (ARP40522_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40522_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ILF3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ILF3. Alternate names: DRBF, DRBP76, MMP4, MPHOSPH4, MPP4, NF-AT-90, NF90, NFAR, NFAR-1, TCP80, CBTF, NF110, NF90a, NF90b, NFAR2, NF110b, TCP110ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-57501