Learn More
Electron Microscopy Sciences Ilford 2000rt Machine Fixer 5 LT
SDP
Catalog No. 5031380
Encompass
2000RT is a non-hardening, liquid fixer concentrate specially designed for the rapid fixing of resin coated paper in processing machines complementing the 2000RT developer. It can be used at a wide range of fixer temperatures, 20 – 40 degree C (68 – 104 degree F). 2000RT Fixer must not be used with fix hardeners.
Catalog No. 50-313-80 Supplier Electron Microscopy Sciences Supplier No. 8306605
2000RT is a non-hardening, liquid fixer concentrate specially designed for the rapid fixing of resin coated paper in processing machines complementing the 2000RT developer. It can be used at a wide range of fixer temperatures, 20 – 40 degree C (68 – 104 degree F). 2000RT Fixer must not be used with fix hardeners.