Learn More
Electron Microscopy Sciences Ilford Pq Universal Developer 5 LT
SDP
Catalog No. 5031376
Encompass
PQ UNIVERSAL is a liquid concentrate dimezones/hydroquinone developer which is suitable for all RC and FB black and white photographic papers. It is used at a dilution of 1+9, it is clean working and it has excellent keeping properties. It gives a slightly warm neutral image tone with most papers.
In addition, PQ UNIVERSAL can be used to dish/tray process ILFORD as well as some other technical films. It is also suitable for dish/tray developing of general purpose sheet films when a fast working, high contrast developer is needed and a high degree of enlargement is not required.
PQ UNIVERSAL developer is designed for use at ambient room temperatures, normally 20 degree C/68 degree F. We do not recommend its use for high temperature or machine processing applications.
Catalog No. 50-313-76 Supplier Electron Microscopy Sciences Supplier No. 8305250
PQ UNIVERSAL is a liquid concentrate dimezones/hydroquinone developer which is suitable for all RC and FB black and white photographic papers. It is used at a dilution of 1+9, it is clean working and it has excellent keeping properties. It gives a slightly warm neutral image tone with most papers.
In addition, PQ UNIVERSAL can be used to dish/tray process ILFORD as well as some other technical films. It is also suitable for dish/tray developing of general purpose sheet films when a fast working, high contrast developer is needed and a high degree of enlargement is not required.
PQ UNIVERSAL developer is designed for use at ambient room temperatures, normally 20 degree C/68 degree F. We do not recommend its use for high temperature or machine processing applications.