Learn More
Electron Microscopy Sciences Ilford Rapid Fixer 1 LT
SDP
Catalog No. 5031378
Encompass
Ilford Rapid Fixer is a non-hardening rapid fixer supplied as a liquid concentrate for fixing black and white film and paper in all manual and machine processing applications. It is easy and convenient to use in the temperature range of 18 – 40 degree C (64 -104 degree F). ILFORD RAPID FIXER must not be used with fix hardeners
Catalog No. 50-313-78 Supplier Electron Microscopy Sciences Supplier No. 8306010
Ilford Rapid Fixer is a non-hardening rapid fixer supplied as a liquid concentrate for fixing black and white film and paper in all manual and machine processing applications. It is easy and convenient to use in the temperature range of 18 – 40 degree C (64 -104 degree F). ILFORD RAPID FIXER must not be used with fix hardeners