Learn More
Electron Microscopy Sciences Ilford Ilfospeed RC Deluxe RC Deluxe Paper, Glossy-Grade 2
SDP
Catalog No. 5031370
Encompass
This is a direct replacement for Kodak Kodabrome II RC B&W Papers. Premium Quality Graded Paper on a Resin Coated Base. MULTIGRADE RC COOLTONE Paper has the standard weight of (190g/m 2) with a resin coated base. It is suitable for a wide range of applications including commercial, press, industrial, advertising, and display work.
Grade: Glossy-Grade 2
Catalog No. 50-313-70 Supplier Electron Microscopy Sciences Supplier No. 8303210
This is a direct replacement for Kodak Kodabrome II RC B&W Papers. Premium Quality Graded Paper on a Resin Coated Base. MULTIGRADE RC COOLTONE Paper has the standard weight of (190g/m 2) with a resin coated base. It is suitable for a wide range of applications including commercial, press, industrial, advertising, and display work.
Grade: Glossy-Grade 2