Learn More
Medchemexpress LLC HY-19631B 5mg Medchemexpress, Ilginatinib hydrochloride CAS:1239358-85-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19631B5MG
Encompass_Preferred
Medchemexpress, HY-19631B 5mg Ilginatinib hydrochloride CAS:1239358-85-0 Ilginatinib hydrochloride (NS-018 hydrochloride) is a highly active and orally bioavailable JAK2 inhibitor, with an IC50 of 0.72 nM, 46-, 54-, and 31-fold selectivity for JAK2 over JAK1 (IC50, 33 nM), JAK3 (IC50, 39 nM), and Tyk2 (IC50, 22 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2155