Learn More
Selleck Chemical LLC Ilginatinib hydrochloride E4630-25MG
SDP
Catalog No. 5000039087
Encompass_Preferred
Ilginatinib hydrochloride (NS-018 hydrochloride) is a potent small-molecule inhibitor of JAK2 that is competitive with adenosine triphosphate (ATP) which exhibits 30 50-fold greater selectivity for JAK2 over other Jak-family kinases such as JAK1 JAK3 and TYK2 It inhibits JAK2 JAK1 JAK3 and TYK2 with IC50 of 0 72 nM 33 nM 39 nM and 22 nM respectively It also showed potent inhibition of Src-family kinases SRC and FYN It can be used in research of myeloproliferative neoplasms
Catalog No. 50-000-39087 Supplier Selleck Chemical LLC Supplier No. E463025MG
Ilginatinib hydrochloride (NS-018 hydrochloride) is a potent small-molecule inhibitor of JAK2 that is competitive with adenosine triphosphate (ATP) which exhibits 30 50-fold greater selectivity for JAK2 over other Jak-family kinases such as JAK1 JAK3 and TYK2 It inhibits JAK2 JAK1 JAK3 and TYK2 with IC50 of 0 72 nM 33 nM 39 nM and 22 nM respectively It also showed potent inhibition of Src-family kinases SRC and FYN It can be used in research of myeloproliferative neoplasms