Learn More
Medchemexpress LLC Ilginatinib hydrochloride | 1239358-85-0 | 98.9% | 425.89 | 25 MG
SDP
Catalog No. 5000402227
Encompass_Preferred
Ilginatinib hydrochloride (NS-018 hydrochloride) is a highly active and orally bioavailable JAK2 inhibitor. It demonstrates significant selectivity for JAK2 over JAK1, JAK3, and Tyk2. This compound is intended for research use only.
  • Highly active and orally bioavailable JAK2 inhibitor.
  • Exhibits selectivity for JAK2 over JAK1, JAK3, and Tyk2.
  • Potently prolongs survival and reduces splenomegaly in mouse models of myeloproliferative neoplasms.
  • Suppresses colony-forming unit-granulocyte/macrophage (CFU-GM) formation from myelodysplastic syndrome (MDS)-derived bone marrow mononuclear cells.
Catalog No. 50-004-02227 Supplier Medchemexpress LLC Supplier No. 25MGHY19631B
Ilginatinib hydrochloride (NS-018 hydrochloride) is a highly active and orally bioavailable JAK2 inhibitor. It demonstrates significant selectivity for JAK2 over JAK1, JAK3, and Tyk2. This compound is intended for research use only.
  • Highly active and orally bioavailable JAK2 inhibitor.
  • Exhibits selectivity for JAK2 over JAK1, JAK3, and Tyk2.
  • Potently prolongs survival and reduces splenomegaly in mouse models of myeloproliferative neoplasms.
  • Suppresses colony-forming unit-granulocyte/macrophage (CFU-GM) formation from myelodysplastic syndrome (MDS)-derived bone marrow mononuclear cells.