Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ilginatinib maleate | 1239358-86-1 | MFCD28502056 | 99.9% | 389.43 | C21H20FN7 | 10 MG
SDP

Catalog No. 5000295356
Encompass_Preferred

Ilginatinib maleate is an orally bioavailable small-molecule JAK2 inhibitor supplied as the maleate salt for laboratory research. It shows sub-nanomolar potency against JAK2 with a selective profile over related kinases, and is provided as a powder with documented solubility and storage guidance for experimental use.

  • Highly potent JAK2 inhibition (IC50 0.72 nM).
  • Demonstrates selectivity versus JAK1, JAK3, and Tyk2.
  • Supplied as a powder suitable for laboratory preparation.
  • High chemical purity (≈99.9%).
  • Soluble in DMSO at 100 mg/mL for stock solutions.
  • Storage and stability data provided for powder and solutions.
  • Intended for research use only; not for clinical or human use.

Catalog No. 50-002-95356 Supplier Medchemexpress LLC Supplier No. HY1963110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ilginatinib maleate is an orally bioavailable small-molecule JAK2 inhibitor supplied as the maleate salt for laboratory research. It shows sub-nanomolar potency against JAK2 with a selective profile over related kinases, and is provided as a powder with documented solubility and storage guidance for experimental use.

  • Highly potent JAK2 inhibition (IC50 0.72 nM).
  • Demonstrates selectivity versus JAK1, JAK3, and Tyk2.
  • Supplied as a powder suitable for laboratory preparation.
  • High chemical purity (≈99.9%).
  • Soluble in DMSO at 100 mg/mL for stock solutions.
  • Storage and stability data provided for powder and solutions.
  • Intended for research use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.