Learn More
Medchemexpress LLC HY-19631 5mg Medchemexpress, Ilginatinib (maleate) CAS:1354799-87-3 Purity:>98%
SDP
Catalog No. 502082153
Encompass_Preferred
Medchemexpress, HY-19631 5mg Ilginatinib (maleate) CAS:1354799-87-3 Ilginatinib maleate (NS-018 maleate) is a highly active and orally bioavailable JAK2 inhibitor, with an IC50 of 0.72 nM, 46-, 54-, and 31-fold selectivity for JAK2 over JAK1 (IC50, 33 nM), JAK3 (IC50, 39 nM), and Tyk2 (IC50, 22 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2153 Supplier Medchemexpress LLC Supplier No. HY196315MG
Medchemexpress, HY-19631 5mg Ilginatinib (maleate) CAS:1354799-87-3 Ilginatinib maleate (NS-018 maleate) is a highly active and orally bioavailable JAK2 inhibitor, with an IC50 of 0.72 nM, 46-, 54-, and 31-fold selectivity for JAK2 over JAK1 (IC50, 33 nM), JAK3 (IC50, 39 nM), and Tyk2 (IC50, 22 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.