Learn More
Cayman Chemical Ilicicolin F, Purity: ≥70%, CAS: 22738-98-3, 1 mg
SDP
Catalog No. 502988785
Encompass_Preferred
A fungal metabolite; inhibits T. vivax alternative oxidase and E. coli cytochrome bo (IC50s = 0.43 and 0.37 µM, respectively) but not E. coli cytochrome bd (IC50 = 85 µM); active against A. fumigatus and C. albicans (MICs = 1.66-3.33 and 6.66-13.33 µg/ml, respectively); cytotoxic to HeLa cells (EC50 = 0.003 µg/ml)
Catalog No. 50-298-8785 Supplier Cayman Chemical Supplier No. 298501 MG
A fungal metabolite; inhibits T. vivax alternative oxidase and E. coli cytochrome bo (IC50s = 0.43 and 0.37 µM, respectively) but not E. coli cytochrome bd (IC50 = 85 µM); active against A. fumigatus and C. albicans (MICs = 1.66-3.33 and 6.66-13.33 µg/ml, respectively); cytotoxic to HeLa cells (EC50 = 0.003 µg/ml)