Learn More
Cayman Chemical IlImaquInon 100ug
SDP
Catalog No. 502969239
Encompass_Preferred
A natural sesquiterpene quinone that reversibly induces vesiculation of Golgi membranes, blocking the secretory pathway; inhibits SAH hydrolase (IC50 = 40 µM), DNA polymerase β (IC50 = 45.2 µM,) and Cdc25B (IC50 = 92 µM); at 10 µM, activates gene expression through HIF-1
Catalog No. 50-296-9239 Supplier Cayman Chemical Supplier No. 15124100 UG
A natural sesquiterpene quinone that reversibly induces vesiculation of Golgi membranes, blocking the secretory pathway; inhibits SAH hydrolase (IC50 = 40 µM), DNA polymerase β (IC50 = 45.2 µM,) and Cdc25B (IC50 = 92 µM); at 10 µM, activates gene expression through HIF-1