Learn More
Medchemexpress LLC ILK-IN-3 10mg | 6975-75-3 | 232.24 | C10H12N6O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11567710MG
Encompass_Preferred
ILK-IN-3 is a small-molecule, orally active inhibitor of integrin-linked kinase (ILK) used for preclinical research. It has been reported to enhance the anticancer efficacy of docetaxel in orthotopic LCC6 models. The compound is supplied as a solid and as a DMSO solution and is provided in milligram pack sizes with high reported purity.
  • Orally active ILK inhibitor for preclinical research
  • Reported to enhance docetaxel efficacy in LCC6 tumor model
  • High reported purity (~99.6%)
  • Available as solid and as 10 mM solution in DMSO
  • Stable under recommended frozen storage conditions
Catalog No. 50-002-26847