Learn More
Medchemexpress LLC ILK-IN-3 25mg | 6975-75-3 | 232.24 g/mol | C10H12N6O | 25 MG
SDP
Catalog No. 5000228753
Encompass_Preferred
ILK-IN-3 is a small-molecule, orally active inhibitor of integrin-linked kinase (ILK) used in preclinical studies to investigate ILK-mediated signaling and evaluate combination cancer therapies. It has been reported to enhance the anticancer efficacy of docetaxel in orthotopic LCC6 models.
  • Orally active small-molecule ILK inhibitor.
  • Shown to potentiate docetaxel efficacy in LCC6 orthotopic model.
  • High chemical purity (99.62%).
  • Molecular formula C10H12N6O; molecular weight ≈ 232.24 g/mol.
  • Supplied in research-scale pack sizes, including 25 mg.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-28753 Supplier Medchemexpress LLC Supplier No. HY11567725MG
ILK-IN-3 is a small-molecule, orally active inhibitor of integrin-linked kinase (ILK) used in preclinical studies to investigate ILK-mediated signaling and evaluate combination cancer therapies. It has been reported to enhance the anticancer efficacy of docetaxel in orthotopic LCC6 models.
  • Orally active small-molecule ILK inhibitor.
  • Shown to potentiate docetaxel efficacy in LCC6 orthotopic model.
  • High chemical purity (99.62%).
  • Molecular formula C10H12N6O; molecular weight ≈ 232.24 g/mol.
  • Supplied in research-scale pack sizes, including 25 mg.
  • Intended for research use only; not for human therapeutic use.