Learn More
Medchemexpress LLC ILK-IN-3 50mg | 6975-75-3 | 50 MG
SDP
Catalog No. 5000226500
Encompass_Preferred
ILK-IN-3 is an orally active integrin-linked kinase (ILK) inhibitor supplied as a 50 mg research reagent. It is used in preclinical studies to modulate ILK activity and has been reported to enhance the anticancer efficacy of docetaxel in orthotopic LCC6 models. For research use only.
  • Orally active ILK inhibitor suitable for in vitro and in vivo studies.
  • Reported to improve docetaxel anticancer efficacy in LCC6 model.
  • High purity (>99% by LCMS) for reproducible results.
  • Supplied as a solid powder with recommended cold storage for stability.
  • Provided with certificate of analysis and datasheet for quality and handling information.
Catalog No. 50-002-26500 Supplier Medchemexpress LLC Supplier No. HY11567750MG
ILK-IN-3 is an orally active integrin-linked kinase (ILK) inhibitor supplied as a 50 mg research reagent. It is used in preclinical studies to modulate ILK activity and has been reported to enhance the anticancer efficacy of docetaxel in orthotopic LCC6 models. For research use only.
  • Orally active ILK inhibitor suitable for in vitro and in vivo studies.
  • Reported to improve docetaxel anticancer efficacy in LCC6 model.
  • High purity (>99% by LCMS) for reproducible results.
  • Supplied as a solid powder with recommended cold storage for stability.
  • Provided with certificate of analysis and datasheet for quality and handling information.