Learn More
Medchemexpress LLC ILK-IN-3 5mg | 6975-75-3 | 5 MG
SDP
Catalog No. 5000228331
Encompass_Preferred
ILK-IN-3 is an orally active small-molecule inhibitor of integrin-linked kinase (ILK) intended for research use. It has formula C10H12N6O and a molecular weight of 232.24 g/mol, and the batch COA reports LCMS purity of 99.62%. It has been reported to enhance the anticancer efficacy of docetaxel in orthotopic LCC6 models.
  • Orally active integrin-linked kinase (ILK) inhibitor
  • High reported purity (99.62% LCMS)
  • Suitable for in vitro and in vivo cancer model studies
  • Available in small mg-scale pack sizes for research (e.g., 5 mg)
  • Provided as a solid with defined molecular formula and weight
Catalog No. 50-002-28331 Supplier Medchemexpress LLC Supplier No. HY1156775MG
ILK-IN-3 is an orally active small-molecule inhibitor of integrin-linked kinase (ILK) intended for research use. It has formula C10H12N6O and a molecular weight of 232.24 g/mol, and the batch COA reports LCMS purity of 99.62%. It has been reported to enhance the anticancer efficacy of docetaxel in orthotopic LCC6 models.
  • Orally active integrin-linked kinase (ILK) inhibitor
  • High reported purity (99.62% LCMS)
  • Suitable for in vitro and in vivo cancer model studies
  • Available in small mg-scale pack sizes for research (e.g., 5 mg)
  • Provided as a solid with defined molecular formula and weight