Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Illudin S | 1149-99-1 | MFCD00870487 | 99.8% | 264.32 | C15H20O4 | 5MG
SDP

Catalog No. 5000239126
Encompass_Preferred

Illudin S is a cytotoxic natural sesquiterpene supplied for research use; it exhibits anti-tumor and antiviral activity and is reported to possess genotoxic properties, including blockage of the G1-S phase transition in human leukemia cells.

  • High purity (99.8%) suitable for analytical and cellular studies.
  • Supplied as a small research quantity (5 mg) in powder form.
  • Solid, white to yellow appearance for straightforward handling.
  • Molecular formula C15H20O4; molecular weight 264.32 g/mol.
  • Documented genotoxic and cell-cycle blocking activity in vitro.
  • Storage guidance: powder -20°C for long term; in solvent -80°C (short term).

Catalog No. 50-002-39126 Supplier Medchemexpress LLC Supplier No. HY1250985MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Illudin S is a cytotoxic natural sesquiterpene supplied for research use; it exhibits anti-tumor and antiviral activity and is reported to possess genotoxic properties, including blockage of the G1-S phase transition in human leukemia cells.

  • High purity (99.8%) suitable for analytical and cellular studies.
  • Supplied as a small research quantity (5 mg) in powder form.
  • Solid, white to yellow appearance for straightforward handling.
  • Molecular formula C15H20O4; molecular weight 264.32 g/mol.
  • Documented genotoxic and cell-cycle blocking activity in vitro.
  • Storage guidance: powder -20°C for long term; in solvent -80°C (short term).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.